The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018462.4 | NP_060932.2 | 2 - 3 | 252 | 72 |
GenBank mRNA | AF220047.1 | - | 1 - 2 | 72 | |
AY148219.1 | - | 2 - 3 | 252 | 72 | |
AY148220.1 | - | 2 - 3 | 252 | 72 | |
BC001067.1 | - | 2 - 3 | 228 | 72 | |
BC007929.1 | - | 2 - 3 | 221 | 72 | |
BC019303.1 | - | 2 - 3 | 219 | 72 | |
BC061515.1 | - | 2 - 3 | 226 | 72 |