The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138392.3 | NP_612401.2 | 16 - 17 | 1813 | 83 |
XM_006723474.1 | XP_006723537.1 | 16 - 17 | 1843 | 83 | |
XM_011527492.1 | XP_011525794.1 | 13 - 14 | 1492 | 83 | |
XM_011527493.1 | XP_011525795.1 | 15 - 16 | 1739 | 83 | |
XM_017027474.1 | XP_016882963.1 | 15 - 16 | 1709 | 83 | |
GenBank mRNA | AF258553.1 | - | 16 - 17 | 1812 | 83 |
AK075057.1 | - | 16 - 17 | 1735 | 83 | |
AK123695.1 | - | 14 - 15 | 1791 | 83 | |
AK297006.1 | - | 10 - 11 | 1439 | 83 | |
AK300819.1 | - | 15 - 16 | 1592 | 83 | |
AK312748.1 | - | 16 - 17 | 1777 | 83 | |
AY007117.1 | - | 7 - 8 | 848 | 83 | |
BC007653.1 | - | 8 - 9 | 1099 | 83 | |
BC022855.1 | - | 16 - 17 | 1784 | 83 | |
CA416700.1 | - | 2 - 3 | 83 |