The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323041.1 | NP_001309970.1 | 3 - 4 | 756 | 88 |
NM_001323042.1 | NP_001309971.1 | 3 - 4 | 240 | 88 | |
NM_001323043.1 | NP_001309972.1 | 3 - 4 | 641 | 88 | |
NM_001323047.1 | NP_001309976.1 | 3 - 4 | 756 | 88 | |
NM_001323048.1 | NP_001309977.1 | 3 - 4 | 641 | 88 | |
NM_001323049.1 | NP_001309978.1 | 2 - 3 | 930 | 88 | |
NM_001323052.1 | NP_001309981.1 | 3 - 4 | 641 | 88 | |
NM_001323053.1 | NP_001309982.1 | 3 - 4 | 756 | 88 | |
NM_006608.2 | NP_006599.2 | 2 - 3 | 545 | 88 | |
NR_136565.1 | 3 - 4 | 641 | 88 | ||
NR_136566.1 | 3 - 4 | 756 | 88 | ||
NR_136567.1 | 3 - 4 | 641 | 88 | ||
GenBank mRNA | AJ011863.1 | - | 2 - 3 | 124 | 88 |
AK294749.1 | - | 2 - 3 | 271 | 88 | |
BC002447.2 | - | 3 - 4 | 689 | 88 | |
CX763183.1 | - | 2 - 3 | 421 | 88 |