The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322253.1 | NP_001309182.1 | 7 - 8 | 989 | 64 |
NM_001322256.1 | NP_001309185.1 | 5 - 6 | 879 | 64 | |
NM_001322257.1 | NP_001309186.1 | 5 - 6 | 708 | 64 | |
NM_015106.3 | NP_055921.2 | 6 - 7 | 941 | 64 | |
XM_006713062.3 | XP_006713125.1 | 5 - 6 | 688 | 64 | |
XM_017005987.1 | XP_016861476.1 | 5 - 6 | 717 | 64 | |
XM_017005988.1 | XP_016861477.1 | 6 - 7 | 2277 | 64 | |
GenBank mRNA | AB018352.2 | - | 3 - 4 | 346 | 64 |
AK122683.1 | - | 5 - 6 | 717 | 64 | |
AK308456.1 | - | 4 - 5 | 478 | 64 | |
BC024298.1 | - | 3 - 4 | 305 | 64 |