The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152641.2 | NP_689854.2 | 8 - 9 | 1019 | 152 |
XM_006719272.3 | XP_006719335.1 | 8 - 9 | 1173 | 152 | |
XM_011538025.1 | XP_011536327.1 | 1 - 2 | 43 | 152 | |
GenBank mRNA | AK055181.1 | - | 1 - 2 | 43 | 152 |
AL832200.1 | - | 4 - 5 | 572 | 152 | |
AY727870.2 | - | 8 - 9 | 1019 | 152 | |
BX648152.1 | - | 1 - 2 | 227 | 152 | |
CR749833.1 | - | 7 - 8 | 938 | 152 | |
DQ096628.1 | - | 8 - 9 | 1019 | 152 |