The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014289.3 | NP_055104.2 | 8 - 9 | 1350 | 70 |
GenBank mRNA | AF029232.1 | - | 8 - 9 | 1250 | 70 |
AJ000388.1 | - | 8 - 9 | 1341 | 70 | |
AK300340.1 | - | 6 - 7 | 906 | 70 | |
AK312701.1 | - | 8 - 9 | 1314 | 70 | |
BC000730.2 | - | 7 - 8 | 1179 | 70 |