The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005505.2 | NP_001005505.1 | 24 - 25 | 2318 | 100 |
NM_001174051.2 | NP_001167522.1 | 25 - 26 | 2339 | 100 | |
NM_001291101.1 | NP_001278030.1 | 24 - 25 | 2114 | 100 | |
NM_006030.3 | NP_006021.2 | 24 - 25 | 2318 | 100 | |
XM_005265581.4 | XP_005265638.1 | 24 - 25 | 2192 | 100 | |
XM_011534243.2 | XP_011532545.1 | 25 - 26 | 2213 | 100 | |
GenBank mRNA | AB011130.1 | - | 24 - 25 | 2145 | 100 |
AF040709.1 | - | 24 - 25 | 2324 | 100 | |
AF042792.1 | - | 25 - 26 | 2305 | 100 | |
AF042793.1 | - | 24 - 25 | 2121 | 100 | |
AJ251367.1 | - | 24 - 25 | 2144 | 100 | |
AJ251368.1 | - | 25 - 26 | 2165 | 100 | |
BC152438.1 | - | 24 - 25 | 2145 | 100 | |
DA760583.1 | - | 7 - 8 | 438 | 100 |