The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005505.2 | NP_001005505.1 | 34 - 35 | 3147 | 114 |
NM_001174051.2 | NP_001167522.1 | 35 - 36 | 3168 | 114 | |
NM_001291101.1 | NP_001278030.1 | 34 - 35 | 2943 | 114 | |
NM_006030.3 | NP_006021.2 | 34 - 35 | 3147 | 114 | |
XM_005265581.4 | XP_005265638.1 | 34 - 35 | 3024 | 114 | |
XM_011534243.2 | XP_011532545.1 | 35 - 36 | 3045 | 114 | |
GenBank mRNA | AB011130.1 | - | 34 - 35 | 2974 | 114 |
AF040709.1 | - | 34 - 35 | 3153 | 114 | |
AF042792.1 | - | 35 - 36 | 3134 | 114 | |
AF042793.1 | - | 34 - 35 | 2950 | 114 | |
AJ251367.1 | - | 34 - 35 | 2973 | 114 | |
AJ251368.1 | - | 35 - 36 | 2994 | 114 | |
AK074820.1 | - | 2 - 3 | 126 | 114 | |
BC152438.1 | - | 34 - 35 | 2974 | 114 |