Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBRM1
Entrez Gene ID
55193
Gene Name
polybromo 1
Gene Aliases
BAF180, PB1
Chromosome Location
Chr.3: 52545352 - 52685933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017006747.1 XP_016862236.1 11 - 12 1768 140
GenBank mRNA AF225870.1 - 8 - 9 896 140

View Full Product Details