The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017647.3 | NP_060117.3 | 3 - 4 | 821 | 74 |
GenBank mRNA | AF327355.1 | - | 2 - 3 | 296 | 74 |
AK095861.1 | - | 3 - 4 | 821 | 74 | |
AK296715.1 | - | 3 - 4 | 220 | 74 | |
AK315010.1 | - | 3 - 4 | 246 | 74 | |
BC036710.2 | - | 2 - 3 | 205 | 74 |