The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017647.3 | NP_060117.3 | 8 - 9 | 1358 | 92 |
GenBank mRNA | AF327355.1 | - | 7 - 8 | 833 | 92 |
AK095861.1 | - | 8 - 9 | 1358 | 92 | |
AK296715.1 | - | 7 - 8 | 564 | 92 | |
AK307398.1 | - | 7 - 8 | 847 | 92 | |
AK315010.1 | - | 8 - 9 | 783 | 92 | |
AL834482.1 | - | 2 - 3 | 246 | 92 | |
BC000131.1 | - | 3 - 4 | 373 | 92 | |
BC036710.2 | - | 7 - 8 | 742 | 92 |