The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000884.2 | NP_000875.2 | 6 - 7 | 713 | 84 |
XM_006713128.3 | XP_006713191.1 | 6 - 7 | 1021 | 84 | |
XM_017006349.1 | XP_016861838.1 | 5 - 6 | 898 | 84 | |
XM_017006350.1 | XP_016861839.1 | 5 - 6 | 898 | 84 | |
GenBank mRNA | AK293397.1 | - | - | 674 | 84 |
AY545560.1 | - | 3 - 4 | 331 | 84 | |
BC006124.1 | - | 6 - 7 | 661 | 84 | |
BC012840.1 | - | 6 - 7 | 654 | 84 | |
BC015567.2 | - | 6 - 7 | 652 | 84 | |
J04208.1 | - | 6 - 7 | 668 | 84 |