The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288962.1 | NP_001275891.1 | 10 - 11 | 1227 | 256 |
NR_110231.1 | 10 - 11 | 1078 | 107 | ||
XM_006722940.1 | XP_006723003.1 | 10 - 11 | 1227 | 256 | |
GenBank mRNA | AB072596.1 | - | 10 - 11 | 1053 | 107 |
AF502289.1 | - | 10 - 11 | 1186 | 256 |