Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDZD4
Entrez Gene ID
57595
Gene Name
PDZ domain containing 4
Gene Aliases
LNX5, LU1, PDZK4, PDZRN4L
Chromosome Location
Chr.X: 153802166 - 153830567 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303512.1 NP_001290441.1 5 - 6 833 81
NM_001303513.1 NP_001290442.1 6 - 7 1076 81
NM_001303514.1 NP_001290443.1 3 - 4 488 81
NM_001303515.1 NP_001290444.1 5 - 6 668 81
NM_001303516.1 NP_001290445.1 5 - 6 650 81
NM_032512.3 NP_115901.2 5 - 6 815 81
GenBank mRNA AB040877.1 - 2 - 3 153 81
AB103478.1 - 5 - 6 796 81
AK091444.1 - 5 - 6 541 81
AK123229.1 - 5 - 6 570 81
AK123822.1 - 5 - 6 668 81
AK127016.1 - 5 - 6 815 81
AK309212.1 - 6 - 7 881 81
BC020347.1 - 4 - 5 378 81
BC117277.1 - 5 - 6 625 81
BC143459.1 - 3 - 4 279 81
BC143545.1 - 5 - 6 624 81

View Full Product Details