Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWA7
Entrez Gene ID
80737
Gene Name
von Willebrand factor A domain containing 7
Gene Aliases
C6orf27, G7c, NG37
Chromosome Location
Chr.6: 31764135 - 31777331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025258.2 NP_079534.2 6 - 7 1151 104
XM_005249427.2 XP_005249484.1 6 - 7 1151 104
XM_017011327.1 XP_016866816.1 5 - 6 1186 104
XM_017011328.1 XP_016866817.1 6 - 7 1297 104
XM_017011329.1 XP_016866818.1 4 - 5 695 104
GenBank mRNA AK304058.1 - 6 - 7 1011 104
AK304266.1 - 5 - 6 924 104
BC114625.1 - 5 - 6 910 104

View Full Product Details