The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270407.1 | NP_001257336.1 | 4 - 5 | 1030 | 88 |
NM_001270408.1 | NP_001257337.1 | 5 - 6 | 1138 | 88 | |
NM_021219.3 | NP_067042.1 | 5 - 6 | 1138 | 88 | |
NR_072999.1 | 5 - 6 | 1138 | 88 | ||
GenBank mRNA | AF255910.1 | - | 5 - 6 | 838 | 88 |
AK294769.1 | - | 4 - 5 | 714 | 88 | |
AK312708.1 | - | 5 - 6 | 910 | 88 | |
AX772824.1 | - | 5 - 6 | 827 | 88 | |
AY016009.1 | - | 5 - 6 | 832 | 88 | |
AY077698.1 | - | 5 - 6 | 880 | 88 | |
AY358361.1 | - | 5 - 6 | 686 | 88 | |
BC017779.1 | - | 5 - 6 | 683 | 88 | |
CX870550.1 | - | 1 - 2 | 122 | 88 |