Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF14
Entrez Gene ID
9678
Gene Name
PHD finger protein 14
Gene Aliases
-
Chromosome Location
Chr.7: 10973834 - 11188565 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014660.3 NP_055475.2 2 - 3 563 117
XM_017012811.1 XP_016868300.1 2 - 3 601 117
XM_017012812.1 XP_016868301.1 2 - 3 601 117
XM_017012813.1 XP_016868302.1 2 - 3 600 117
XM_017012814.1 XP_016868303.1 2 - 3 600 117
XM_017012815.1 XP_016868304.1 2 - 3 600 117
XM_017012816.1 XP_016868305.1 2 - 3 599 117
GenBank mRNA AB018326.1 - 2 - 3 518 117
BC152414.1 - 2 - 3 518 117

View Full Product Details