The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102371.1 | NP_001095841.1 | 3 - 4 | 874 | 55 |
NM_024955.5 | NP_079231.4 | 3 - 4 | 913 | 55 | |
GenBank mRNA | AK025714.1 | - | 3 - 4 | 813 | 55 |
AK225347.1 | - | 3 - 4 | 813 | 55 | |
AK315555.1 | - | 3 - 4 | 816 | 55 | |
BC015726.1 | - | 3 - 4 | 797 | 55 | |
BC027716.2 | - | 3 - 4 | 892 | 55 | |
BC040351.1 | - | 3 - 4 | 836 | 55 | |
CR456444.1 | - | 2 - 3 | 888 | 55 |