Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APTX
Entrez Gene ID
54840
Gene Name
aprataxin
Gene Aliases
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
Chromosome Location
Chr.9: 32883872 - 33025129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017014833.1 XP_016870322.1 7 - 8 947 82

View Full Product Details