The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005373.3 | NP_001005373.1 | 16 - 17 | 1536 | 83 |
NM_001005374.3 | NP_001005374.1 | 15 - 16 | 1241 | 83 | |
NM_001190723.2 | NP_001177652.1 | 16 - 17 | 1536 | 83 | |
NM_138361.5 | NP_612370.3 | 15 - 16 | 1795 | 83 | |
XM_006717316.3 | XP_006717379.1 | 16 - 17 | 1536 | 83 | |
XM_017015283.1 | XP_016870772.1 | 16 - 17 | 2535 | 83 | |
XM_017015284.1 | XP_016870773.1 | 16 - 17 | 1531 | 83 | |
GenBank mRNA | AK056203.1 | - | 16 - 17 | 83 | |
AK056305.1 | - | 4 - 5 | 381 | 83 | |
AK091589.1 | - | 15 - 16 | 1543 | 83 | |
AK226106.1 | - | 16 - 17 | 1522 | 83 | |
AY358830.1 | - | 16 - 17 | 1501 | 83 | |
BC009239.2 | - | 15 - 16 | 1241 | 83 |