Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRSAM1
Entrez Gene ID
90678
Gene Name
leucine rich repeat and sterile alpha motif containing 1
Gene Aliases
CMT2P, RIFLE, TAL
Chromosome Location
Chr.9: 127451486 - 127503501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005373.3 NP_001005373.1 24 - 25 2287 73
NM_001005374.3 NP_001005374.1 23 - 24 1992 73
NM_001190723.2 NP_001177652.1 23 - 24 2206 73
NM_138361.5 NP_612370.3 23 - 24 2546 73
XM_006717316.3 XP_006717379.1 23 - 24 2188 73
XM_017015283.1 XP_016870772.1 24 - 25 3286 73
XM_017015284.1 XP_016870773.1 24 - 25 2282 73
GenBank mRNA AK056203.1 - 23 - 24 2188 73
AK056305.1 - 12 - 13 1137 73
AK091589.1 - 23 - 24 2294 73
AK097094.1 - 4 - 5 1747 73
AK226106.1 - 24 - 25 2273 73
AY358830.1 - 24 - 25 2252 73
BC009239.2 - 23 - 24 1992 73

View Full Product Details