The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005373.3 | NP_001005373.1 | 3 - 4 | 445 | 107 |
NM_001005374.3 | NP_001005374.1 | 2 - 3 | 150 | 107 | |
NM_001190723.2 | NP_001177652.1 | 3 - 4 | 445 | 107 | |
NM_138361.5 | NP_612370.3 | 2 - 3 | 704 | 107 | |
XM_006717316.3 | XP_006717379.1 | 3 - 4 | 445 | 107 | |
XM_017015283.1 | XP_016870772.1 | 3 - 4 | 1444 | 107 | |
XM_017015284.1 | XP_016870773.1 | 3 - 4 | 445 | 107 | |
GenBank mRNA | AK056203.1 | - | 3 - 4 | 427 | 107 |
AK091589.1 | - | 2 - 3 | 452 | 107 | |
AK226106.1 | - | 3 - 4 | 431 | 107 | |
AY358830.1 | - | 3 - 4 | 410 | 107 | |
BC009239.2 | - | 2 - 3 | 150 | 107 | |
DB165274.1 | - | 3 - 4 | 445 | 107 |