Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC16B
Entrez Gene ID
89866
Gene Name
SEC16 homolog B, endoplasmic reticulum export factor
Gene Aliases
LZTR2, PGPR-p117, RGPR, SEC16S
Chromosome Location
Chr.1: 177929107 - 177969915 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033127.2 NP_149118.2 4 - 5 622 72
GenBank mRNA AB063357.1 - 4 - 5 622 72
AK314585.1 - 4 - 5 72
BC009106.1 - 4 - 5 614 72
EF125213.1 - 3 - 4 532 72

View Full Product Details