The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080522.2 | NP_001073991.2 | 4 - 5 | 375 | 84 |
NM_001164720.1 | NP_001158192.1 | 3 - 4 | 341 | 84 | |
XM_005248177.1 | XP_005248234.1 | 3 - 4 | 330 | 84 | |
XM_011513872.2 | XP_011512174.1 | 3 - 4 | 330 | 84 | |
XM_011513874.2 | XP_011512176.1 | 4 - 5 | 1580 | 84 | |
GenBank mRNA | AK096047.1 | - | 3 - 4 | 317 | 84 |
AK309178.1 | - | 3 - 4 | 310 | 84 | |
BC053865.1 | - | 3 - 4 | 294 | 84 | |
BX647334.1 | - | 3 - 4 | 310 | 84 | |
DA860072.1 | - | 3 - 4 | 330 | 84 | |
EU450799.1 | - | 4 - 5 | 202 | 84 | |
N35522.1 | - | 3 - 4 | 341 | 84 |