Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHG3
Entrez Gene ID
26030
Gene Name
pleckstrin homology and RhoGEF domain containing G3
Gene Aliases
ARHGEF43, KIAA0599
Chromosome Location
Chr.14: 64704129 - 64744346 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308147.1 NP_001295076.1 8 - 9 1366 130
XM_011536624.2 XP_011534926.1 8 - 9 1194 130
XM_011536625.2 XP_011534927.1 8 - 9 1154 130
XM_011536627.2 XP_011534929.1 8 - 9 1459 130
XM_011536628.2 XP_011534930.1 8 - 9 1645 130
XM_011536629.2 XP_011534931.1 8 - 9 1146 130
XM_017021154.1 XP_016876643.1 8 - 9 1150 130
XM_017021155.1 XP_016876644.1 8 - 9 1149 130
XM_017021156.1 XP_016876645.1 8 - 9 1217 130
XM_017021157.1 XP_016876646.1 8 - 9 1466 130
XM_017021158.1 XP_016876647.1 6 - 7 652 130
GenBank mRNA BC129953.1 - 8 - 9 1177 130
BX248779.1 - 6 - 7 1170 130

View Full Product Details