The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308147.1 | NP_001295076.1 | 5 - 6 | 896 | 93 |
XM_011536624.2 | XP_011534926.1 | 5 - 6 | 724 | 93 | |
XM_011536625.2 | XP_011534927.1 | 5 - 6 | 684 | 93 | |
XM_011536627.2 | XP_011534929.1 | 5 - 6 | 989 | 93 | |
XM_011536628.2 | XP_011534930.1 | 5 - 6 | 1175 | 93 | |
XM_011536629.2 | XP_011534931.1 | 5 - 6 | 676 | 93 | |
XM_017021154.1 | XP_016876643.1 | 5 - 6 | 680 | 93 | |
XM_017021155.1 | XP_016876644.1 | 5 - 6 | 679 | 93 | |
XM_017021156.1 | XP_016876645.1 | 5 - 6 | 747 | 93 | |
XM_017021157.1 | XP_016876646.1 | 5 - 6 | 996 | 93 | |
XM_017021158.1 | XP_016876647.1 | 3 - 4 | 182 | 93 | |
GenBank mRNA | BC129952.1 | - | 5 - 6 | 707 | 93 |
BC129953.1 | - | 5 - 6 | 707 | 93 |