The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020133.2 | NP_064518.1 | 9 - 9 | 3980 | 70 |
XM_005267053.3 | XP_005267110.1 | 8 - 8 | 3796 | 70 | |
XM_006715514.3 | XP_006715577.1 | 8 - 8 | 3804 | 70 | |
XM_017011059.1 | XP_016866548.1 | 10 - 10 | 4150 | 70 | |
GenBank mRNA | AL122123.1 | - | 1 - 1 | 1373 | 70 |
CR627395.1 | - | 8 - 8 | 3757 | 70 |