Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- USP28
- Entrez Gene ID
-
57646
- Gene Name
- ubiquitin specific peptidase 28
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 113797875 - 113875580 on Build GRCh38
View Full Product Details