The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306144.1 | NP_001293073.1 | 3 - 4 | 442 | 77 |
GenBank mRNA | AK226109.1 | - | 3 - 4 | 396 | 77 |
AK295738.1 | - | 3 - 4 | 237 | 77 |