The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018702.3 | NP_061172.1 | 1 - 2 | 421 | 106 |
GenBank mRNA | AF034837.1 | - | 1 - 2 | 429 | 106 |
AI695657.1 | - | 2 - 3 | 106 | ||
BC137477.1 | - | 1 - 2 | 242 | 106 | |
BC140852.1 | - | 1 - 2 | 242 | 106 |