The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018702.3 | NP_061172.1 | 3 - 4 | 1403 | 100 |
GenBank mRNA | AF034837.1 | - | 3 - 4 | 1411 | 100 |
BC137477.1 | - | 3 - 4 | 1224 | 100 | |
BC140852.1 | - | 3 - 4 | 1224 | 100 |