The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018702.3 | NP_061172.1 | 9 - 10 | 2372 | 90 |
GenBank mRNA | AF034837.1 | - | 9 - 10 | 2380 | 90 |
AK055107.1 | - | 2 - 3 | 577 | 90 | |
AK057763.1 | - | 2 - 3 | 275 | 90 | |
AK289895.1 | - | 2 - 3 | 609 | 90 | |
AK307451.1 | - | 2 - 3 | 442 | 90 | |
AK308579.1 | - | 3 - 4 | 347 | 90 | |
BC047443.1 | - | 2 - 3 | 246 | 90 | |
BC137477.1 | - | 9 - 10 | 2193 | 90 | |
BC139725.1 | - | 2 - 3 | 253 | 90 | |
BC140852.1 | - | 9 - 10 | 2193 | 90 |