The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014494.2 | NP_055309.2 | 23 - 24 | 5492 | 120 |
XM_005255254.3 | XP_005255311.1 | 22 - 23 | 5335 | 120 | |
XM_005255257.4 | XP_005255314.1 | 22 - 23 | 5459 | 120 | |
XM_017023143.1 | XP_016878632.1 | 22 - 23 | 5436 | 120 | |
XM_017023144.1 | XP_016878633.1 | 21 - 22 | 5403 | 120 | |
XM_017023145.1 | XP_016878634.1 | 22 - 23 | 5369 | 120 | |
XM_017023146.1 | XP_016878635.1 | 22 - 23 | 5501 | 120 | |
XM_017023147.1 | XP_016878636.1 | 22 - 23 | 5529 | 120 | |
XM_017023148.1 | XP_016878637.1 | 21 - 22 | 5256 | 120 | |
XM_017023149.1 | XP_016878638.1 | 20 - 21 | 5253 | 120 | |
XM_017023150.1 | XP_016878639.1 | 21 - 22 | 5218 | 120 | |
XM_017023151.1 | XP_016878640.1 | 20 - 21 | 5107 | 120 | |
XM_017023152.1 | XP_016878641.1 | 21 - 22 | 5008 | 120 | |
XM_017023153.1 | XP_016878642.1 | 22 - 23 | 5459 | 120 | |
XM_017023154.1 | XP_016878643.1 | 20 - 21 | 5350 | 120 | |
GenBank mRNA | AB040893.1 | - | 18 - 19 | 3692 | 120 |
AJ492221.1 | - | 22 - 23 | 5378 | 120 | |
AK001773.1 | - | 6 - 7 | 817 | 120 | |
AK025696.1 | - | 16 - 17 | 2049 | 120 | |
AL833757.1 | - | 17 - 18 | 2653 | 120 | |
AY035864.1 | - | 20 - 21 | 5026 | 120 | |
BC024324.1 | - | 6 - 7 | 838 | 120 | |
BC068209.1 | - | 1 - 2 | 363 | 120 |