Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNRC6A
Entrez Gene ID
27327
Gene Name
trinucleotide repeat containing 6A
Gene Aliases
CAGH26, GW1, GW182, TNRC6
Chromosome Location
Chr.16: 24610260 - 24826227 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014494.2 NP_055309.2 9 - 10 3674 76
XM_005255254.3 XP_005255311.1 9 - 10 3664 76
XM_005255257.4 XP_005255314.1 8 - 9 3641 76
XM_017023143.1 XP_016878632.1 8 - 9 3618 76
XM_017023145.1 XP_016878634.1 8 - 9 3620 76
XM_017023146.1 XP_016878635.1 8 - 9 3683 76
XM_017023147.1 XP_016878636.1 9 - 10 3858 76
XM_017023148.1 XP_016878637.1 7 - 8 3438 76
XM_017023150.1 XP_016878639.1 8 - 9 3616 76
XM_017023151.1 XP_016878640.1 7 - 8 3436 76
XM_017023152.1 XP_016878641.1 7 - 8 3190 76
XM_017023153.1 XP_016878642.1 8 - 9 3641 76
GenBank mRNA AB040893.1 - 4 - 5 1874 76
AJ492221.1 - 8 - 9 3560 76
AK025696.1 - 3 - 4 378 76
AL833757.1 - 4 - 5 982 76
AY035864.1 - 6 - 7 3208 76

View Full Product Details