The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198963.1 | NP_945314.1 | 22 - 23 | 3922 | 137 |
XM_011533154.2 | XP_011531456.1 | 22 - 23 | 3926 | 137 | |
XM_011533155.2 | XP_011531457.1 | 22 - 23 | 3924 | 137 | |
XM_011533156.2 | XP_011531458.1 | 22 - 23 | 3926 | 137 | |
GenBank mRNA | AF070590.1 | - | 7 - 8 | 935 | 137 |
AF283512.1 | - | 18 - 19 | 2613 | 137 | |
BC033636.1 | - | 3 - 4 | 292 | 137 | |
BC039270.1 | - | 21 - 22 | 3752 | 137 | |
BC053623.1 | - | 16 - 17 | 2223 | 137 | |
BC065278.1 | - | 17 - 18 | 2417 | 137 | |
BC131534.1 | - | 22 - 23 | 3894 | 137 |