The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198963.1 | NP_945314.1 | 8 - 9 | 1998 | 68 |
XM_011533154.2 | XP_011531456.1 | 8 - 9 | 2002 | 68 | |
XM_011533155.2 | XP_011531457.1 | 8 - 9 | 2000 | 68 | |
XM_011533156.2 | XP_011531458.1 | 8 - 9 | 2002 | 68 | |
XM_011533157.1 | XP_011531459.1 | 8 - 9 | 2002 | 68 | |
GenBank mRNA | AF283512.1 | - | 4 - 5 | 689 | 68 |
AK296739.1 | - | 8 - 9 | 2037 | 68 | |
AK307514.1 | - | 8 - 9 | 2027 | 68 | |
BC039270.1 | - | 7 - 8 | 1828 | 68 | |
BC053623.1 | - | 2 - 3 | 299 | 68 | |
BC065278.1 | - | 3 - 4 | 493 | 68 | |
BC131534.1 | - | 8 - 9 | 1970 | 68 | |
BU183594.1 | - | 3 - 4 | 492 | 68 |