The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001312686.1 | NP_001299615.1 | 11 - 12 | 1589 | 196 |
NM_014813.2 | NP_055628.1 | 10 - 11 | 1477 | 196 | |
XM_005271369.2 | XP_005271426.1 | 10 - 11 | 1477 | 196 | |
GenBank mRNA | AB018349.1 | - | 10 - 11 | 1438 | 196 |
AK291031.1 | - | 10 - 11 | 1469 | 196 | |
BC117368.1 | - | 10 - 11 | 1301 | 196 | |
BC117370.1 | - | 10 - 11 | 1301 | 196 |