The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004066.2 | NP_004057.1 | 1 - 1 | 119 | |
GenBank mRNA | AI198628.1 | - | 1 - 1 | 129 | 119 |