The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022819.3 | NP_073730.3 | 1 - 2 | 210 | 110 |
XM_011541955.1 | XP_011540257.1 | 1 - 2 | 203 | 110 | |
XM_011541956.1 | XP_011540258.1 | 1 - 2 | 214 | 110 | |
GenBank mRNA | AK093645.1 | - | 1 - 2 | 214 | 110 |
BX647458.1 | - | 1 - 2 | 210 | 110 |