The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020886.3 | NP_065937.1 | 18 - 19 | 2371 | 75 |
XM_005271637.2 | XP_005271694.1 | 18 - 19 | 2370 | 75 | |
XM_017018057.1 | XP_016873546.1 | 19 - 20 | 2697 | 75 | |
XM_017018058.1 | XP_016873547.1 | 17 - 18 | 2303 | 75 | |
XM_017018061.1 | XP_016873550.1 | 16 - 17 | 2163 | 75 | |
XM_017018062.1 | XP_016873551.1 | 16 - 17 | 2160 | 75 | |
XM_017018064.1 | XP_016873553.1 | 16 - 17 | 2168 | 75 | |
XM_017018065.1 | XP_016873554.1 | 19 - 20 | 2412 | 75 | |
XM_017018066.1 | XP_016873555.1 | 20 - 21 | 2774 | 75 | |
XM_017018067.1 | XP_016873556.1 | 15 - 16 | 2023 | 75 | |
GenBank mRNA | AF266283.1 | - | 18 - 19 | 2308 | 75 |