The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270768.1 | NP_001257697.1 | 14 - 15 | 1450 | 96 |
NM_033088.3 | NP_149079.2 | 14 - 15 | 1603 | 96 | |
NR_073071.1 | 15 - 16 | 1717 | 96 | ||
XM_006710995.2 | XP_006711058.1 | 7 - 8 | 706 | 96 | |
XM_017002629.1 | XP_016858118.1 | 14 - 15 | 1602 | 96 | |
GenBank mRNA | AB051548.2 | - | 14 - 15 | 1568 | 96 |
AK027649.1 | - | 14 - 15 | 1450 | 96 | |
AK125054.1 | - | 7 - 8 | 1372 | 96 | |
AK291059.1 | - | 14 - 15 | 1577 | 96 | |
AL834196.1 | - | 15 - 16 | 1686 | 96 | |
BC094786.1 | - | 14 - 15 | 1584 | 96 | |
BC119814.1 | - | 14 - 15 | 1585 | 96 | |
BC121793.1 | - | 14 - 15 | 1585 | 96 |