The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003750.2 | NP_003741.1 | 3 - 4 | 524 | 138 |
GenBank mRNA | AK302575.1 | - | 3 - 4 | 469 | 138 |
AK307610.1 | - | 3 - 4 | 502 | 138 | |
AU279600.1 | - | 3 - 4 | 524 | 138 | |
BC033769.1 | - | 3 - 4 | 497 | 138 | |
BC054342.1 | - | 3 - 4 | 483 | 138 | |
BC064926.1 | - | 3 - 4 | 508 | 138 | |
BC107776.1 | - | 3 - 4 | 516 | 138 | |
BC114429.1 | - | 3 - 4 | 432 | 138 | |
D50929.1 | - | 3 - 4 | 506 | 138 | |
JX312508.1 | - | 3 - 4 | 525 | 138 | |
U58046.1 | - | 3 - 4 | 476 | 138 | |
U78311.1 | - | 3 - 4 | 491 | 138 |