The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139283.1 | NP_644812.1 | 5 - 6 | 1078 | 83 |
GenBank mRNA | AF385435.1 | - | 5 - 6 | 1078 | 83 |
AK124744.1 | - | 6 - 7 | 922 | 83 | |
AY357944.1 | - | 5 - 6 | 847 | 83 | |
BC111551.1 | - | 6 - 7 | 880 | 83 |