Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWA7
Entrez Gene ID
80737
Gene Name
von Willebrand factor A domain containing 7
Gene Aliases
C6orf27, G7c, NG37
Chromosome Location
Chr.6: 31764135 - 31777331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025258.2 NP_079534.2 1 - 2 220 106
XM_005249427.2 XP_005249484.1 1 - 2 220 106
GenBank mRNA AK304058.1 - 1 - 2 196 106
DC323829.1 - 1 - 2 221 106

View Full Product Details