Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WWC3
Entrez Gene ID
55841
Gene Name
WWC family member 3
Gene Aliases
BM042
Chromosome Location
Chr.X: 10015755 - 10144478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AI306162.1 - 1 - 2 47 92

View Full Product Details