The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015691.3 | NP_056506.2 | 7 - 8 | 861 | 79 |
GenBank mRNA | AB033106.1 | - | 5 - 6 | 484 | 79 |
AK292961.1 | - | 6 - 7 | 743 | 79 | |
AL353937.1 | - | 3 - 4 | 341 | 79 | |
BC032447.1 | - | 4 - 5 | 396 | 79 | |
KC987947.1 | - | 7 - 8 | 1038 | 79 |