Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEKT5
Entrez Gene ID
146279
Gene Name
tektin 5
Gene Aliases
CT149
Chromosome Location
Chr.16: 10584610 - 10742416 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144674.1 NP_653275.1 6 - 7 1314 58
XM_011522383.2 XP_011520685.1 6 - 7 1314 58
XM_011522385.2 XP_011520687.1 6 - 7 1314 58
XM_011522389.1 XP_011520691.1 6 - 7 1314 58
XM_011522390.2 XP_011520692.1 9 - 10 1671 58
XM_017022955.1 XP_016878444.1 6 - 7 1314 58
XM_017022956.1 XP_016878445.1 6 - 7 1211 58
XM_017022958.1 XP_016878447.1 3 - 4 578 58
GenBank mRNA AK057433.1 - 6 - 7 1314 58
BC130336.1 - 6 - 7 1297 58
BC130338.1 - 6 - 7 1297 58

View Full Product Details