The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000439.4 | NP_000430.3 | 2 - 3 | 527 | 132 |
NM_001177875.1 | NP_001171346.1 | 2 - 3 | 185 | 132 | |
GenBank mRNA | AB208874.1 | - | 2 - 3 | 203 | 132 |
AK303888.1 | - | 2 - 3 | 185 | 132 | |
BC130295.1 | - | 2 - 3 | 409 | 132 | |
BC136486.1 | - | 2 - 3 | 409 | 132 | |
M90753.1 | - | 2 - 3 | 497 | 132 | |
X64810.1 | - | 2 - 3 | 477 | 132 |