The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006912.5 | NP_008843.1 | 1 - 2 | 158 | 101 |
GenBank mRNA | AF084462.1 | - | 1 - 2 | 69 | 101 |
AK314239.1 | - | 1 - 2 | 102 | ||
BM546985.1 | - | 1 - 2 | 152 | 101 | |
U71203.1 | - | 1 - 2 | 90 | 101 | |
U78165.1 | - | 1 - 2 | 90 | 101 | |
Y07566.1 | - | 1 - 2 | 100 | 101 |