Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PBRM1
Entrez Gene ID
55193
Gene Name
polybromo 1
Gene Aliases
BAF180, PB1
Chromosome Location
Chr.3: 52545352 - 52685933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011533900.2 XP_011532202.1 3 - 4 363 76
XM_011533902.2 XP_011532204.1 2 - 3 295 76
XM_011533903.2 XP_011532205.1 2 - 3 300 76
XM_017006725.1 XP_016862214.1 2 - 3 243 76
XM_017006726.1 XP_016862215.1 3 - 4 353 76
XM_017006727.1 XP_016862216.1 3 - 4 298 76
XM_017006728.1 XP_016862217.1 2 - 3 294 76
XM_017006729.1 XP_016862218.1 2 - 3 702 76
XM_017006730.1 XP_016862219.1 3 - 4 298 76
XM_017006731.1 XP_016862220.1 2 - 3 281 76
XM_017006732.1 XP_016862221.1 2 - 3 295 76
XM_017006733.1 XP_016862222.1 2 - 3 701 76
XM_017006734.1 XP_016862223.1 2 - 3 274 76
XM_017006735.1 XP_016862224.1 2 - 3 701 76
XM_017006736.1 XP_016862225.1 2 - 3 702 76
XM_017006741.1 XP_016862230.1 2 - 3 277 76
XM_017006742.1 XP_016862231.1 2 - 3 300 76
XM_017006743.1 XP_016862232.1 2 - 3 702 76
XM_017006744.1 XP_016862233.1 2 - 3 275 76
XM_017006747.1 XP_016862236.1 2 - 3 703 76
XM_017006751.1 XP_016862240.1 2 - 3 277 76
XM_017006752.1 XP_016862241.1 2 - 3 702 76
XM_017006753.1 XP_016862242.1 2 - 3 701 76
XM_017006754.1 XP_016862243.1 2 - 3 295 76
XM_017006755.1 XP_016862244.1 2 - 3 277 76
XM_017006756.1 XP_016862245.1 2 - 3 277 76
XM_017006759.1 XP_016862248.1 2 - 3 277 76
XM_017006760.1 XP_016862249.1 2 - 3 295 76
XM_017006761.1 XP_016862250.1 2 - 3 304 76
XM_017006762.1 XP_016862251.1 2 - 3 295 76
XM_017006763.1 XP_016862252.1 3 - 4 363 76
XM_017006764.1 XP_016862253.1 2 - 3 277 76
XM_017006766.1 XP_016862255.1 2 - 3 284 76

View Full Product Details